3-Bromo-4-oxo-4-phenylbutanoic acid ≥95%

Supplier: Matrix Scientific

044480-500MG 044480-1G 044480-5G
101866-092EA 181.54 USD
101866-092 101866-094 101866-096
3-Bromo-4-oxo-4-phenylbutanoic acid ≥95%
3-Bromo-4-oxo-4-phenylbutanoic acid

Hazard: Irritant

Formula: C₁₀H₉BrO₃
Melting Pt: 121…124 °C
MDL Number: MFCD00086369
CAS Number: 53515-22-3

Order Now

Specification Test Results

Melting point 121-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR