1-Methyl-3-phenyl-5-(phenylsulfanyl)-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
044476-500MG
044476-1G
044476-5G
101866-068EA
747.69
USD
101866-068
101866-070
101866-072
1-Methyl-3-phenyl-5-(phenylsulfanyl)-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-3-phenyl-5-(phenylsulfanyl)-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₇H₁₄N₂OS Melting Pt: 81…84 °C |
MDL Number:
MFCD00244933 CAS Number: 318247-51-7 |
Specification Test Results
Melting point | 81-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...