5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044475-500MG 044475-1G 044475-5G
101866-062EA 747.69 USD
101866-062 101866-064 101866-066
5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₃ClN₂OS
Melting Pt: 146…149 °C
MDL Number: MFCD00244931
CAS Number: 318247-49-3

Order Now

Specification Test Results

Melting point 146-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR