4-Nitro-3-[(phenylsulfonyl)methyl]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044474-500MG 044474-1G 044474-5G
101866-056EA 421.54 USD
101866-056 101866-058 101866-060
4-Nitro-3-[(phenylsulfonyl)methyl]benzenecarbaldehyde ≥95%
4-Nitro-3-[(phenylsulfonyl)methyl]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁NO₅S
MW: 305.31 g/mol
Melting Pt: 200…202 °C
MDL Number: MFCD00794650
CAS Number: 339276-48-1

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR