1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

044473-500MG 044473-1G 044473-5G
101866-050EA 892.31 USD
101866-050 101866-052 101866-054
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxylic acid ≥95%
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O₂
Melting Pt: 200-202°C
MDL Number: MFCD00664523
CAS Number: 339276-36-7

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR