Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044472-500MG 044472-1G 044472-5G
101866-044EA 78.46 USD
101866-044 101866-046 101866-048
Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate ≥95%
Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃O₂
MW: 255.71 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD00664522
CAS Number: 339276-34-5

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR