2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide ≥95%

Supplier: Matrix Scientific

044469-500MG 044469-1G 044469-5G
101866-026EA 375.38 USD
101866-026 101866-028 101866-030
2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide ≥95%
2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₀F₃NO₂
MW: 245.2 g/mol
Melting Pt: 62…66 °C
MDL Number: MFCD00232083
CAS Number: 338424-38-7

Order Now

Specification Test Results

Melting point 62-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR