N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide ≥95%
Supplier: Matrix Scientific
044468-500MG
044468-1G
044468-5G
101866-020EA
375.38
USD
101866-020
101866-022
101866-024
N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide ≥95%
N-(4-Chlorophenyl)-2-methyl-2-oxiranecarboxamide
Hazard: Irritant
Formula:
C₁₀H₁₀ClNO₂ Melting Pt: 90…94 °C |
MDL Number:
MFCD00232082 CAS Number: 65925-79-3 |
Specification Test Results
Melting point | 90-94°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...