4-Chloro-N-(2-pyrimidinyl)butanamide ≥95%

Supplier: Matrix Scientific

044465-500MG 044465-1G 044465-5G
101866-006EA 181.54 USD
101866-006 101866-008 101866-010
4-Chloro-N-(2-pyrimidinyl)butanamide ≥95%
4-Chloro-N-(2-pyrimidinyl)butanamide

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD00664561
CAS Number: 27179-31-3

Order Now

Specification Test Results

Melting point 100-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR