2-[5-(4-Chlorobenzoyl)-2-thienyl]acetonitrile ≥95%

Supplier: Matrix Scientific

044464-500MG 044464-1G 044464-5G
101866-000EA 278.46 USD
101866-000 101866-002 101866-004
2-[5-(4-Chlorobenzoyl)-2-thienyl]acetonitrile ≥95%
2-[5-(4-Chlorobenzoyl)-2-thienyl]acetonitrile

Hazard: Irritant

Formula: C₁₃H₈ClNOS
Melting Pt: 148…150 °C
MDL Number: MFCD00664560
CAS Number: 338422-77-8

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR