2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetohydrazide ≥95%

Supplier: Matrix Scientific

044463-500MG 044463-1G 044463-5G
101865-994EA 747.69 USD
101865-994 101865-996 101865-998
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetohydrazide ≥95%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetohydrazide

Hazard: Irritant

Formula: C₈H₇ClF₃N₃OS
Melting Pt: 150 °C
MDL Number: MFCD00664272
CAS Number: 338422-76-7

Order Now

Specification Test Results

Melting point 150°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR