Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-3-(dimethylamino)acrylate ≥95%

Supplier: Matrix Scientific

044462-500MG 044462-1G 044462-5G
101865-988EA 747.69 USD
101865-988 101865-990 101865-992
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-3-(dimethylamino)acrylate ≥95%
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-3-(dimethylamino)acrylate

Hazard: Irritant

Formula: C₁₃H₁₄ClF₃N₂O₂S
MW: 354.78 g/mol
Melting Pt: 102…105 °C
MDL Number: MFCD00664271
CAS Number: 338422-75-6

Order Now

Specification Test Results

Melting point 102-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR