2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetic acid ≥95%

Supplier: Matrix Scientific

044461-500MG 044461-1G 044461-5G
101865-982EA 92.31 USD
101865-982 101865-984 101865-986
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetic acid ≥95%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetic acid

Hazard: Irritant

Formula: C₈H₅ClF₃NO₂S
Melting Pt: 124…127 °C
MDL Number: MFCD00245200
CAS Number: 338422-73-4

Order Now

Specification Test Results

Melting point 124-127°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR