2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid ≥95%

Supplier: Matrix Scientific

044459-500MG 044459-1G 044459-5G
101865-970EA 747.69 USD
101865-970 101865-972 101865-974
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid ≥95%
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid

Hazard: Irritant

Formula: C₁₁H₉ClF₃N₃O₄
MW: 339.66 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD00173606
CAS Number: 338422-72-3

Order Now

Specification Test Results

Melting point 222-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR