4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

044458-500MG 044458-1G 044458-5G
101865-964EA 375.38 USD
101865-964 101865-966 101865-968
4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride ≥95%
4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₂H₆Cl₂F₃NO₃S
MW: 372.139 g/mol
Melting Pt: 69…72 °C
MDL Number: MFCD00243963
CAS Number: 338422-71-2

Order Now

Specification Test Results

Melting point 69-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR