Methyl-1-[(3,4-dichloroanilino)carbonyl]-4-hydroxy-2-pyrrolidinecarboxylate ≥95%
Supplier: Matrix Scientific
044457-500MG
044457-1G
044457-5G
101865-958EA
921.54
USD
101865-958
101865-960
101865-962
Methyl-1-[(3,4-dichloroanilino)carbonyl]-4-hydroxy-2-pyrrolidinecarboxylate ≥95%
Methyl-1-[(3,4-dichloroanilino)carbonyl]-4-hydroxy-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₄Cl₂N₂O₄ Melting Pt: 175…177 °C |
MDL Number:
MFCD00202783 CAS Number: 318247-31-3 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...