Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate ≥95%

Supplier: Matrix Scientific

044456-500MG 044456-1G 044456-5G
101865-952EA 921.54 USD
101865-952 101865-954 101865-956
Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate ≥95%
Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₅F₃N₂O₄
MW: 332.28 g/mol
Melting Pt: 183…185 °C
MDL Number: MFCD00202782
CAS Number: 318247-30-2

Order Now

Specification Test Results

Melting point 183-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR