1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulfanyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Fluoro-2-[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-ylthio]acetophenone

044453-500MG 044453-1G 044453-5G
101865-934EA 747.69 USD
101865-934 101865-936 101865-938
1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulfanyl}-1-ethanone ≥95%
1-(4-Fluorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulfanyl}-1-ethanone

Hazard: Irritant

Formula: C₁₃H₁₃FN₂O₂S
MW: 280.32 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00215160
CAS Number: 338422-62-1

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR