(2-{[2-(4-Methoxyphenoxy)ethyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%

Supplier: Matrix Scientific

044452-500MG 044452-1G 044452-5G
101865-928EA 747.69 USD
101865-928 101865-930 101865-932
(2-{[2-(4-Methoxyphenoxy)ethyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%
(2-{[2-(4-Methoxyphenoxy)ethyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃S
Melting Pt: 124…126 °C
MDL Number: MFCD00215158
CAS Number: 338422-59-6

Order Now

Specification Test Results

Melting point 124-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR