(2-{[(5-Chloro-1,2,3-thiadiazol-4-yl)methyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%
Supplier: Matrix Scientific
044448-500MG
044448-1G
044448-5G
101865-904EA
1109.23
USD
101865-904
101865-906
101865-908
(2-{[(5-Chloro-1,2,3-thiadiazol-4-yl)methyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%
(2-{[(5-Chloro-1,2,3-thiadiazol-4-yl)methyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol
Hazard: Irritant
Formula:
C₈H₉ClN₄OS₂ Melting Pt: 131…133 °C |
MDL Number:
MFCD00232307 CAS Number: 338422-47-2 |
Specification Test Results
Melting point | 131-133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...