(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}-5-(hydroxymethyl)-1-methyl-1H-imidazole

044447-500MG 044447-1G 044447-5G
101865-898EA 747.69 USD
101865-898 101865-900 101865-902
(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol ≥95%
(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol

Hazard: Irritant

Formula: C₁₁H₉ClF₃N₃OS
Melting Pt: 93…95 °C
MDL Number: MFCD00215151
CAS Number: 338422-44-9

Order Now

Specification Test Results

Melting point 93-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR