5-(Methylamino)-3-morpholino-4-isothiazolecarbonitrile ≥95%

Supplier: Matrix Scientific

044445-500MG 044445-1G 044445-5G
101865-886EA 1483.08 USD
101865-886 101865-888 101865-890
5-(Methylamino)-3-morpholino-4-isothiazolecarbonitrile ≥95%
5-(Methylamino)-3-morpholino-4-isothiazolecarbonitrile

Hazard: Irritant

Formula: C₉H₁₂N₄OS
Melting Pt: 176…178 °C
MDL Number: MFCD00243960
CAS Number: 338422-41-6

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR