{2-[(4-Fluorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol ≥95%

Supplier: Matrix Scientific

044444-500MG 044444-1G 044444-5G
101865-880EA 375.38 USD
101865-880 101865-882 101865-884
{2-[(4-Fluorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol ≥95%
{2-[(4-Fluorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol

Hazard: Irritant

Formula: C₁₂H₁₃FN₂OS
Melting Pt: 124…126 °C
MDL Number: MFCD00215148
CAS Number: 338422-40-5

Order Now

Specification Test Results

Melting point 124-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR