2-({2-[(4-Methylbenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

044441-500MG 044441-1G 044441-5G
101865-862EA 181.54 USD
101865-862 101865-864 101865-866
2-({2-[(4-Methylbenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%
2-({2-[(4-Methylbenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃S
MW: 253.32 g/mol
Melting Pt: 131…133 °C
MDL Number: MFCD00202396
CAS Number: 338421-46-8

Order Now

Specification Test Results

Melting point 131-133°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR