2-({2-[(4-Chlorobenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

044439-500MG 044439-1G 044439-5G
101865-850EA 181.54 USD
101865-850 101865-852 101865-854
2-({2-[(4-Chlorobenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid ≥95%
2-({2-[(4-Chlorobenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃S
Melting Pt: 144…147 °C
MDL Number: MFCD00202308
CAS Number: 338421-25-3

Order Now

Specification Test Results

Melting point 144-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR