2-{[2-(2-Chloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%

Supplier: Matrix Scientific

044438-500MG 044438-1G 044438-5G
101865-844EA 181.54 USD
101865-844 101865-846 101865-848
2-{[2-(2-Chloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%
2-{[2-(2-Chloroanilino)-2-oxoethyl]sulfanyl}acetic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃S
Melting Pt: 128…130 °C
MDL Number: MFCD00105057
CAS Number: 338421-17-3

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR