2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Thiodiacetic acid mono-3-(trifluoromethyl)anilide

044436-500MG 044436-1G 044436-5G
101865-832EA 181.54 USD
101865-832 101865-834 101865-836
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid ≥95%
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₀F₃NO₃S
MW: 293.27 g/mol
Melting Pt: 102…104 °C
MDL Number: MFCD00202305
CAS Number: 338421-05-9

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR