Methyl-1-methyl-2-sulfanyl-1H-imidazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-mercapto-1-methyl-1H-imidazole-5-carboxylate

044435-500MG 044435-1G 044435-5G
101865-826EA 747.69 USD
101865-826 101865-828 101865-830
Methyl-1-methyl-2-sulfanyl-1H-imidazole-5-carboxylate ≥95%
Methyl-1-methyl-2-sulfanyl-1H-imidazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 205…207 °C
MDL Number: MFCD00206762
CAS Number: 68892-07-9

Order Now

Specification Test Results

Melting point 205-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR