2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

044434-500MG 044434-1G 044434-5G
101865-820EA 181.54 USD
101865-820 101865-822 101865-824
2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile ≥95%
2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₅H₉FN₂
Melting Pt: 87…89 °C
MDL Number: MFCD00231953
CAS Number: 116617-31-3

Order Now

Specification Test Results

Melting point 87-89°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR