Methyl-4-[(1-formyl-2-naphthyl)oxy]-2-butenoate ≥95%

Supplier: Matrix Scientific

044433-500MG 044433-1G 044433-5G
101865-814EA 375.38 USD
101865-814 101865-816 101865-818
Methyl-4-[(1-formyl-2-naphthyl)oxy]-2-butenoate ≥95%
Methyl-4-[(1-formyl-2-naphthyl)oxy]-2-butenoate

Hazard: Irritant

Formula: C₁₆H₁₄O₄
Melting Pt: 98…101 °C
MDL Number: MFCD00231679
CAS Number: 174633-84-2

Order Now

Specification Test Results

Melting point 98-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR