3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Dimethylamino)-1-(4-nitrophenyl)prop-2-en-1-one

044432-500MG 044432-1G 044432-5G
101865-808EA 181.54 USD
101865-808 101865-810 101865-812
3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one ≥97%
3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 151…154 °C
MDL Number: MFCD00121191
CAS Number: 68760-11-2

Order Now

Specification Test Results

Melting point 151-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR