Ethyl-4-(4-fluorobenzoyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044430-500MG 044430-1G
101865-798EA 461.54 USD
101865-798 101865-800
Ethyl-4-(4-fluorobenzoyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(4-fluorobenzoyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀FNO₄
MW: 263.23 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD00664758
CAS Number: 338420-60-3

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR