1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

Synonyms: N-(3-Trifluoromethyl)phenylmaleimide

044429-500MG 044429-1G 044429-5G
101865-792EA 146.15 USD
101865-792 101865-794 101865-796
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione ≥95%
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₁H₆F₃NO₂
Melting Pt: 62…65 °C
MDL Number: MFCD00439448
CAS Number: 53629-19-9

Order Now

Specification Test Results

Melting point 62-65°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR