[5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%

Supplier: Matrix Scientific

044426-500MG 044426-1G 044426-5G
101865-776EA 747.69 USD
101865-776 101865-778 101865-780
[5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₂H₁₀ClF₃N₂O₂
Melting Pt: 83…85 °C
MDL Number: MFCD00172650
CAS Number: 318239-67-7

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR