6-Methoxy-2H-chromene-3-carbonitrile ≥95%
Supplier: Matrix Scientific
044425-500MG
044425-1G
044425-5G
101865-770EA
278.46
USD
101865-770
101865-772
101865-774
6-Methoxy-2H-chromene-3-carbonitrile ≥95%
6-Methoxy-2H-chromene-3-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₉NO₂ Melting Pt: 75 °C |
MDL Number:
MFCD00202032 CAS Number: 57543-71-2 |
Specification Test Results
Melting point | 75°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...