1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine

Supplier: Matrix Scientific

002853-1G 002853-5G
101784-762EA 36.92 USD
101784-762 101784-764
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine

Hazard: Irritant/Refrigerate

Formula: C₈H₈F₃N₃O₂
Melting Pt: 68…70 °C
MDL Number: MFCD00052691
CAS Number: 120940-43-4

Order Now

Specification Test Results

Melting point 68-70°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR