3-Fluoro-DL-valine ≥95%

Supplier: Matrix Scientific

002820-100MG 002820-250MG
101784-714EA 158.46 USD
101784-714 101784-716
3-Fluoro-DL-valine ≥95%
3-Fluoro-DL-valine

Hazard: Irritant

Formula: C₅H₁₀FNO₂
MW: 135.14 g/mol
Melting Pt: ∼ 250 °C (dec.)
MDL Number: MFCD00077549
CAS Number: 43163-94-6

Order Now

Specification Test Results

Melting point ca 250°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR