4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

002792-1G 002792-5G
101784-672EA 112.31 USD
101784-672 101784-674
4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline ≥97%
4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₁H₆ClF₃N₂O₂
MW: 290.63 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00126373
CAS Number: 175203-61-9

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR