2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
Supplier: Matrix Scientific
002747-1G
002747-5G
101784-568EA
72.31
USD
101784-568
101784-570
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
2-Fluoro-6-(4-(methylthio)phenoxy)benzonitrile
Hazard: Irritant
Formula:
C₁₄H₁₀FNOS Melting Pt: 68…71 °C |
MDL Number:
MFCD00068069 CAS Number: 148901-52-4 |
Specification Test Results
Melting point | 68-71°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...