2-Fluoro-6-(4-methylphenoxy)benzonitrile
Supplier: Matrix Scientific
002733-1G
002733-5G
101784-542EA
52.31
USD
101784-542
101784-544
2-Fluoro-6-(4-methylphenoxy)benzonitrile
2-Fluoro-6-(4-methylphenoxy)benzonitrile
Hazard: Irritant
Formula:
C₁₄H₁₀FNO Melting Pt: 93…95 °C |
MDL Number:
MFCD00068203 CAS Number: 175204-08-7 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...