2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile

Supplier: Matrix Scientific

002729-250MG 002729-1G
101784-534EA 43.08 USD
101784-534 101784-536
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile

Hazard: Irritant

Formula: C₁₄H₉ClFNO
Melting Pt: 146…148 °C
MDL Number: MFCD00068205
CAS Number: 175204-10-1

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR