Methyl-2,5-bis(trifluoroethoxy)benzoate ≥95%

Supplier: Matrix Scientific

002679-1G 002679-5G
101784-416EA 46.15 USD
101784-416 101784-418
Methyl-2,5-bis(trifluoroethoxy)benzoate ≥95%
Methyl-2,5-bis(trifluoroethoxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₀F₆O₄
MW: 332.2 g/mol
Melting Pt: 40…41 °C
MDL Number: MFCD00236693
CAS Number: 35480-31-0

Order Now

Specification Test Results

Melting point 40-41°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR