3-Methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylic acid hydrazide

Supplier: Matrix Scientific

002678-250MG 002678-1G
101784-412EA 44.62 USD
101784-412 101784-414
3-Methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylic acid hydrazide
3-Methyl-5-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxylic acid hydrazide

Hazard: Irritant

Formula: C₁₂H₁₀F₃N₃O₂
Melting Pt: 155…158 °C
MDL Number: MFCD00102588
CAS Number: 175276-90-1

Order Now

Specification Test Results

Melting point 155-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR