5-Methyl-4-phenyl-o-anisidine ≥98%

Supplier: Matrix Scientific

002600-1G 002600-5G
101784-264EA 67.69 USD
101784-264 101784-266
5-Methyl-4-phenyl-o-anisidine ≥98%
5-Methyl-4-phenyl-o-anisidine

Hazard: Irritant/Irritant-Harmful

Formula: C₁₄H₁₅NO
Melting Pt: 65-67°C
MDL Number: MFCD00270117
CAS Number: 206761-76-4

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR