3-Ethoxy-4-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

002561-5G 002561-25G
101784-164EA 153.85 USD
101784-164 101784-166
3-Ethoxy-4-methoxyphenylacetonitrile ≥98%
3-Ethoxy-4-methoxyphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₁H₁₃NO₂
Melting Pt: 58…60 °C
CAS Number: 103796-99-2

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR