5-Fluoro-4-hydroxyquinazoline

Supplier: Matrix Scientific

002541-250MG 002541-1G 002541-5G
101784-110EA 15.38 USD
101784-110 101784-112 101784-114
5-Fluoro-4-hydroxyquinazoline
5-Fluoro-4-hydroxyquinazoline

Hazard: Irritant

Formula: C₈H₅FN₂O
MW: 164.14 g/mol
Melting Pt: 240…242 °C
MDL Number: MFCD20542997
CAS Number: 436-72-6

Order Now

Specification Test Results

Melting point 240-242°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR