3-Ethoxy-4-hydroxyphenylacetonitrile ≥98%
Supplier: Matrix Scientific
002540-5G
002540-25G
101784-106EA
123.08
USD
101784-106
101784-108
3-Ethoxy-4-hydroxyphenylacetonitrile ≥98%
3-Ethoxy-4-hydroxyphenylacetonitrile
Hazard: Irritant/Irritant-Harmful
Formula:
C₁₀H₁₁NO₂ Melting Pt: 92-94°C |
CAS Number:
205748-01-2 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...