3-Chloro-4-fluorophenylhydrazine hydrochloride

Supplier: Matrix Scientific

Synonyms: (3-Chloro-4-fluorophenyl)hydrazine hydrochloride

002517-25G 002517-100G
101784-034EA 46.15 USD
101784-034 101784-036
3-Chloro-4-fluorophenylhydrazine hydrochloride
3-Chloro-4-fluorophenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂FN₂
MW: 197.04 g/mol
Melting Pt: 211-212°C
MDL Number: MFCD00052267
CAS Number: 175135-74-7

Order Now

Specification Test Results

Melting point 211-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR