6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine
Supplier: Matrix Scientific
002447-250MG
002447-1G
101783-940EA
30.77
USD
101783-940
101783-942
6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine
6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine
Hazard: Irritant
Formula:
C₇H₈F₃N₃ MW: 191.16 g/mol Melting Pt: 82…83 °C |
MDL Number:
MFCD00052901 CAS Number: 22123-09-7 |
Specification Test Results
Melting point | 82-83°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...