4-Ethoxy-3-methoxyphenylacetic acid

Supplier: Matrix Scientific

002364-5G 002364-25G
101783-766EA 115.38 USD
101783-766 101783-768
4-Ethoxy-3-methoxyphenylacetic acid
4-Ethoxy-3-methoxyphenylacetic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MW: 210.2300 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00016831
CAS Number: 120-13-8

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR